Assay Details
| Gene Symbol: Cish | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834157.2 | XP_004834214.1 | 1 - 2 | 145 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004834157.2 transcripts.
Set Membership: |
|

Cish
Genomic Map