Assay Details
| Gene Symbol: LOC101715399 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838650.2 | XP_004838707.1 | 8 - 9 | 1242 | 98 | |
| XM_021253187.1 | XP_021108846.1 | 8 - 9 | 1067 | 98 | ||
More Information
Set Membership: |
|

LOC101715399
Genomic Map