Assay Details
| Gene Symbol: LOC101713282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839017.3 | XP_004839074.3 | 9 - 10 | 1728 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101713282| Gene Symbol: LOC101713282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839017.3 | XP_004839074.3 | 9 - 10 | 1728 | 76 | |
Set Membership: |
|