Assay Details
| Gene Symbol: LOC101717251 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839793.3 | XP_004839850.1 | 23 - 24 | 3257 | 119 | |
| XM_004839795.3 | XP_004839852.2 | 23 - 24 | 3147 | 119 | ||
| XM_004839796.3 | XP_004839853.2 | 22 - 23 | 2883 | 119 | ||
| XM_013070363.2 | XP_012925817.1 | 24 - 25 | 3340 | 119 | ||
| XM_013070364.2 | XP_012925818.1 | 21 - 22 | 2695 | 119 | ||
| XM_013070365.2 | XP_012925819.1 | 20 - 21 | 2772 | 119 | ||
| XM_013070367.2 | XP_012925821.1 | 14 - 15 | 1855 | 119 | ||
| XM_013070369.2 | XP_012925823.1 | 6 - 7 | 876 | 119 | ||
| XM_021254020.1 | XP_021109679.1 | 18 - 19 | 2382 | 119 | ||
| XM_021254021.1 | XP_021109680.1 | 5 - 6 | 747 | 119 | ||
More Information
Set Membership: |
|

LOC101717251
Genomic Map