Assay Details
| Gene Symbol: LOC101717251 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839793.3 | XP_004839850.1 | 3 - 4 | 464 | 73 | |
| XM_004839795.3 | XP_004839852.2 | 3 - 4 | 465 | 73 | ||
| XM_013070363.2 | XP_012925817.1 | 3 - 4 | 466 | 73 | ||
| XM_013070366.2 | XP_012925820.1 | 3 - 4 | 473 | 73 | ||
More Information
Set Membership: |
|

LOC101717251
Genomic Map