Assay Details
| Gene Symbol: Ovol2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840862.3 | XP_004840919.1 | 3 - 4 | 575 | 64 | |
| XM_013070855.2 | XP_012926309.1 | 2 - 3 | 274 | 64 | ||
| XM_013070856.2 | XP_012926310.1 | 2 - 3 | 287 | 64 | ||
| XM_013070857.2 | XP_012926311.1 | 2 - 3 | 333 | 64 | ||
| XM_021255105.1 | XP_021110764.1 | 2 - 3 | 1130 | 64 | ||
More Information
Set Membership: |
|

Ovol2
Genomic Map