Assay Details
| Gene Symbol: Selplg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843627.3 | XP_004843684.1 | 2 - 3 | 221 | 69 | |
| XM_021257322.1 | XP_021112981.1 | 2 - 3 | 474 | 69 | ||
| XM_021257324.1 | XP_021112983.1 | 2 - 3 | 317 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004843627.3, XM_021257324.1 transcripts.
Set Membership: |
|

Selplg
Genomic Map