Assay Details
| Gene Symbol: LOC101721703 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848051.3 | XP_004848108.1 | 14 - 15 | 2306 | 105 | |
| XM_021261053.1 | XP_021116712.1 | 13 - 14 | 1729 | 105 | ||
More Information
Set Membership: |
|

LOC101721703
Genomic Map