Assay Details
| Gene Symbol: LOC101721703 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848051.3 | XP_004848108.1 | 1 - 2 | 556 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004848051.3 transcripts.
Set Membership: |
|

LOC101721703
Genomic Map