Assay Details
| Gene Symbol: LOC101721703 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848051.3 | XP_004848108.1 | 8 - 9 | 1512 | 125 | |
| XM_021261053.1 | XP_021116712.1 | 7 - 8 | 935 | 125 | ||
More Information
Set Membership: |
|

LOC101721703
Genomic Map