Assay Details
| Gene Symbol: G6pc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857617.2 | XP_004857674.1 | 2 - 3 | 354 | 64 | |
| XM_004857618.2 | XP_004857675.1 | 2 - 3 | 354 | 64 | ||
More Information
Set Membership: |
|

G6pc2
Genomic Map