Assay Details
Gene Symbol: Syce2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004872402.3 | XP_004872459.1 | 1 - 2 | 1262 | 65 | |
XM_021250363.1 | XP_021106022.1 | 1 - 2 | 1737 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004872402.3, XM_021250363.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
