Assay Details
| Gene Symbol: LOC101716530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004875488.3 | XP_004875545.1 | 11 - 12 | 1694 | 93 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101716530| Gene Symbol: LOC101716530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004875488.3 | XP_004875545.1 | 11 - 12 | 1694 | 93 | |