Assay Details
| Gene Symbol: LOC101716511 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021251257.1 | XP_021106916.1 | 1 - 2 | 569 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101716511| Gene Symbol: LOC101716511 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021251257.1 | XP_021106916.1 | 1 - 2 | 569 | 102 | |
Set Membership: |
|