Assay Details
| Gene Symbol: LOC101714008 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021255540.1 | XP_021111199.1 | 5 - 6 | 577 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101714008| Gene Symbol: LOC101714008 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021255540.1 | XP_021111199.1 | 5 - 6 | 577 | 89 | |
Set Membership: |
|