Assay Details
| Gene Symbol: Pkia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021256004.1 | XP_021111663.1 | 1 - 2 | 160 | 69 | |
| XM_021256005.1 | XP_021111664.1 | 1 - 2 | 444 | 69 | ||
More Information
Set Membership: |
|

Pkia
Genomic Map