Assay Details
| Gene Symbol: LOC101724144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840404.2 | XP_004840461.1 | 10 - 11 | 2468 | 77 | |
| XM_021254663.1 | XP_021110322.1 | 16 - 17 | 4213 | 77 | ||
| XM_021254664.1 | XP_021110323.1 | 14 - 15 | 3421 | 77 | ||
| XM_021254665.1 | XP_021110324.1 | 14 - 15 | 3598 | 77 | ||
| XM_021254666.1 | XP_021110325.1 | 14 - 15 | 3283 | 77 | ||
| XM_021254667.1 | XP_021110326.1 | 14 - 15 | 2945 | 77 | ||
| XM_021254668.1 | XP_021110327.1 | 15 - 16 | 4141 | 77 | ||
| XM_021254669.1 | XP_021110328.1 | 15 - 16 | 4138 | 77 | ||
| XM_021254670.1 | XP_021110329.1 | 13 - 14 | 3346 | 77 | ||
| XM_021254671.1 | XP_021110330.1 | 15 - 16 | 3461 | 77 | ||
| XM_021254672.1 | XP_021110331.1 | 16 - 17 | 4181 | 77 | ||
| XM_021254673.1 | XP_021110332.1 | 15 - 16 | 2875 | 77 | ||
| XM_021254674.1 | XP_021110333.1 | 14 - 15 | 2818 | 77 | ||
| XM_021254675.1 | XP_021110334.1 | 10 - 11 | 2519 | 77 | ||
| XM_021254676.1 | XP_021110335.1 | 13 - 14 | 2778 | 77 | ||

LOC101724144
Genomic Map