Assay Details
| Gene Symbol: LOC101724144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840404.2 | XP_004840461.1 | 11 - 12 | 2663 | 58 | |
| XM_021254663.1 | XP_021110322.1 | 17 - 18 | 4408 | 58 | ||
| XM_021254664.1 | XP_021110323.1 | 15 - 16 | 3616 | 58 | ||
| XM_021254665.1 | XP_021110324.1 | 15 - 16 | 3793 | 58 | ||
| XM_021254666.1 | XP_021110325.1 | 15 - 16 | 3478 | 58 | ||
| XM_021254667.1 | XP_021110326.1 | 15 - 16 | 3140 | 58 | ||
| XM_021254668.1 | XP_021110327.1 | 16 - 17 | 4336 | 58 | ||
| XM_021254669.1 | XP_021110328.1 | 16 - 17 | 4333 | 58 | ||
| XM_021254670.1 | XP_021110329.1 | 14 - 15 | 3541 | 58 | ||
| XM_021254671.1 | XP_021110330.1 | 16 - 17 | 3656 | 58 | ||
| XM_021254672.1 | XP_021110331.1 | 17 - 18 | 4376 | 58 | ||
| XM_021254673.1 | XP_021110332.1 | 16 - 17 | 3070 | 58 | ||
| XM_021254674.1 | XP_021110333.1 | 15 - 16 | 3013 | 58 | ||
| XM_021254675.1 | XP_021110334.1 | 11 - 12 | 2714 | 58 | ||
| XM_021254676.1 | XP_021110335.1 | 14 - 15 | 2973 | 58 | ||
More Information
Set Membership: |
|

LOC101724144
Genomic Map