Assay Details
| Gene Symbol: LOC101724144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840404.2 | XP_004840461.1 | 12 - 13 | 3017 | 109 | |
| XM_021254663.1 | XP_021110322.1 | 18 - 19 | 4762 | 109 | ||
| XM_021254664.1 | XP_021110323.1 | 16 - 17 | 3970 | 109 | ||
| XM_021254665.1 | XP_021110324.1 | 16 - 17 | 4147 | 109 | ||
| XM_021254666.1 | XP_021110325.1 | 16 - 17 | 3832 | 109 | ||
| XM_021254667.1 | XP_021110326.1 | 16 - 17 | 3494 | 109 | ||
| XM_021254668.1 | XP_021110327.1 | 17 - 18 | 4690 | 109 | ||
| XM_021254669.1 | XP_021110328.1 | 17 - 18 | 4687 | 109 | ||
| XM_021254670.1 | XP_021110329.1 | 15 - 16 | 3895 | 109 | ||
| XM_021254671.1 | XP_021110330.1 | 17 - 18 | 4010 | 109 | ||
| XM_021254672.1 | XP_021110331.1 | 18 - 19 | 4730 | 109 | ||
| XM_021254673.1 | XP_021110332.1 | 17 - 18 | 3424 | 109 | ||
| XM_021254674.1 | XP_021110333.1 | 16 - 17 | 3367 | 109 | ||
| XM_021254675.1 | XP_021110334.1 | 12 - 13 | 3068 | 109 | ||
| XM_021254676.1 | XP_021110335.1 | 15 - 16 | 3327 | 109 | ||
More Information
Set Membership: |
|

LOC101724144
Genomic Map