Assay Details
| Gene Symbol: LOC101724144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840404.2 | XP_004840461.1 | 13 - 14 | 3154 | 91 | |
| XM_021254663.1 | XP_021110322.1 | 19 - 20 | 4899 | 91 | ||
| XM_021254664.1 | XP_021110323.1 | 17 - 18 | 4107 | 91 | ||
| XM_021254665.1 | XP_021110324.1 | 17 - 18 | 4284 | 91 | ||
| XM_021254666.1 | XP_021110325.1 | 17 - 18 | 3969 | 91 | ||
| XM_021254667.1 | XP_021110326.1 | 17 - 18 | 3631 | 91 | ||
| XM_021254668.1 | XP_021110327.1 | 18 - 19 | 4827 | 91 | ||
| XM_021254669.1 | XP_021110328.1 | 18 - 19 | 4824 | 91 | ||
| XM_021254670.1 | XP_021110329.1 | 16 - 17 | 4032 | 91 | ||
| XM_021254671.1 | XP_021110330.1 | 18 - 19 | 4147 | 91 | ||
| XM_021254672.1 | XP_021110331.1 | 19 - 20 | 4867 | 91 | ||
| XM_021254673.1 | XP_021110332.1 | 18 - 19 | 3561 | 91 | ||
| XM_021254674.1 | XP_021110333.1 | 17 - 18 | 3504 | 91 | ||
| XM_021254675.1 | XP_021110334.1 | 13 - 14 | 3205 | 91 | ||
| XM_021254676.1 | XP_021110335.1 | 16 - 17 | 3464 | 91 | ||

LOC101724144
Genomic Map