Assay Details
| Gene Symbol: LOC101724144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840404.2 | XP_004840461.1 | 14 - 15 | 3288 | 72 | |
| XM_021254663.1 | XP_021110322.1 | 20 - 21 | 5033 | 72 | ||
| XM_021254664.1 | XP_021110323.1 | 18 - 19 | 4241 | 72 | ||
| XM_021254665.1 | XP_021110324.1 | 18 - 19 | 4418 | 72 | ||
| XM_021254666.1 | XP_021110325.1 | 18 - 19 | 4103 | 72 | ||
| XM_021254667.1 | XP_021110326.1 | 18 - 19 | 3765 | 72 | ||
| XM_021254668.1 | XP_021110327.1 | 19 - 20 | 4961 | 72 | ||
| XM_021254669.1 | XP_021110328.1 | 19 - 20 | 4958 | 72 | ||
| XM_021254670.1 | XP_021110329.1 | 17 - 18 | 4166 | 72 | ||
| XM_021254671.1 | XP_021110330.1 | 19 - 20 | 4281 | 72 | ||
| XM_021254672.1 | XP_021110331.1 | 20 - 21 | 5001 | 72 | ||
| XM_021254673.1 | XP_021110332.1 | 19 - 20 | 3695 | 72 | ||
| XM_021254674.1 | XP_021110333.1 | 18 - 19 | 3638 | 72 | ||
| XM_021254675.1 | XP_021110334.1 | 14 - 15 | 3339 | 72 | ||
| XM_021254676.1 | XP_021110335.1 | 17 - 18 | 3598 | 72 | ||
More Information
Set Membership: |
|

LOC101724144
Genomic Map