Assay Details
| Gene Symbol: LOC101724144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840404.2 | XP_004840461.1 | 2 - 3 | 721 | 94 | |
| XM_021254663.1 | XP_021110322.1 | 8 - 9 | 2466 | 94 | ||
| XM_021254664.1 | XP_021110323.1 | 6 - 7 | 1674 | 94 | ||
| XM_021254665.1 | XP_021110324.1 | 6 - 7 | 1851 | 94 | ||
| XM_021254666.1 | XP_021110325.1 | 6 - 7 | 1536 | 94 | ||
| XM_021254667.1 | XP_021110326.1 | 6 - 7 | 1198 | 94 | ||
| XM_021254668.1 | XP_021110327.1 | 7 - 8 | 2394 | 94 | ||
| XM_021254669.1 | XP_021110328.1 | 7 - 8 | 2391 | 94 | ||
| XM_021254670.1 | XP_021110329.1 | 5 - 6 | 1599 | 94 | ||
| XM_021254671.1 | XP_021110330.1 | 7 - 8 | 1714 | 94 | ||
| XM_021254672.1 | XP_021110331.1 | 8 - 9 | 2434 | 94 | ||
| XM_021254673.1 | XP_021110332.1 | 7 - 8 | 1128 | 94 | ||
| XM_021254674.1 | XP_021110333.1 | 6 - 7 | 1071 | 94 | ||
| XM_021254675.1 | XP_021110334.1 | 2 - 3 | 772 | 94 | ||
| XM_021254676.1 | XP_021110335.1 | 5 - 6 | 1031 | 94 | ||

LOC101724144
Genomic Map