Assay Details
| Gene Symbol: LOC101724144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840404.2 | XP_004840461.1 | 3 - 4 | 964 | 64 | |
| XM_021254663.1 | XP_021110322.1 | 9 - 10 | 2709 | 64 | ||
| XM_021254664.1 | XP_021110323.1 | 7 - 8 | 1917 | 64 | ||
| XM_021254665.1 | XP_021110324.1 | 7 - 8 | 2094 | 64 | ||
| XM_021254666.1 | XP_021110325.1 | 7 - 8 | 1779 | 64 | ||
| XM_021254667.1 | XP_021110326.1 | 7 - 8 | 1441 | 64 | ||
| XM_021254668.1 | XP_021110327.1 | 8 - 9 | 2637 | 64 | ||
| XM_021254669.1 | XP_021110328.1 | 8 - 9 | 2634 | 64 | ||
| XM_021254670.1 | XP_021110329.1 | 6 - 7 | 1842 | 64 | ||
| XM_021254671.1 | XP_021110330.1 | 8 - 9 | 1957 | 64 | ||
| XM_021254672.1 | XP_021110331.1 | 9 - 10 | 2677 | 64 | ||
| XM_021254673.1 | XP_021110332.1 | 8 - 9 | 1371 | 64 | ||
| XM_021254674.1 | XP_021110333.1 | 7 - 8 | 1314 | 64 | ||
| XM_021254675.1 | XP_021110334.1 | 3 - 4 | 1015 | 64 | ||
| XM_021254676.1 | XP_021110335.1 | 6 - 7 | 1274 | 64 | ||
More Information
Set Membership: |
|

LOC101724144
Genomic Map