Assay Details
| Gene Symbol: LOC101724144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840404.2 | XP_004840461.1 | 4 - 5 | 1596 | 68 | |
| XM_021254663.1 | XP_021110322.1 | 10 - 11 | 3341 | 68 | ||
| XM_021254664.1 | XP_021110323.1 | 8 - 9 | 2549 | 68 | ||
| XM_021254665.1 | XP_021110324.1 | 8 - 9 | 2726 | 68 | ||
| XM_021254666.1 | XP_021110325.1 | 8 - 9 | 2411 | 68 | ||
| XM_021254667.1 | XP_021110326.1 | 8 - 9 | 2073 | 68 | ||
| XM_021254668.1 | XP_021110327.1 | 9 - 10 | 3269 | 68 | ||
| XM_021254669.1 | XP_021110328.1 | 9 - 10 | 3266 | 68 | ||
| XM_021254670.1 | XP_021110329.1 | 7 - 8 | 2474 | 68 | ||
| XM_021254671.1 | XP_021110330.1 | 9 - 10 | 2589 | 68 | ||
| XM_021254672.1 | XP_021110331.1 | 10 - 11 | 3309 | 68 | ||
| XM_021254673.1 | XP_021110332.1 | 9 - 10 | 2003 | 68 | ||
| XM_021254674.1 | XP_021110333.1 | 8 - 9 | 1946 | 68 | ||
| XM_021254675.1 | XP_021110334.1 | 4 - 5 | 1647 | 68 | ||
| XM_021254676.1 | XP_021110335.1 | 7 - 8 | 1906 | 68 | ||
More Information
Set Membership: |
|

LOC101724144
Genomic Map