Assay Details
| Gene Symbol: LOC101724144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840404.2 | XP_004840461.1 | 7 - 8 | 1907 | 86 | |
| XM_021254663.1 | XP_021110322.1 | 13 - 14 | 3652 | 86 | ||
| XM_021254664.1 | XP_021110323.1 | 11 - 12 | 2860 | 86 | ||
| XM_021254665.1 | XP_021110324.1 | 11 - 12 | 3037 | 86 | ||
| XM_021254666.1 | XP_021110325.1 | 11 - 12 | 2722 | 86 | ||
| XM_021254667.1 | XP_021110326.1 | 11 - 12 | 2384 | 86 | ||
| XM_021254668.1 | XP_021110327.1 | 12 - 13 | 3580 | 86 | ||
| XM_021254669.1 | XP_021110328.1 | 12 - 13 | 3577 | 86 | ||
| XM_021254670.1 | XP_021110329.1 | 10 - 11 | 2785 | 86 | ||
| XM_021254671.1 | XP_021110330.1 | 12 - 13 | 2900 | 86 | ||
| XM_021254672.1 | XP_021110331.1 | 13 - 14 | 3620 | 86 | ||
| XM_021254673.1 | XP_021110332.1 | 12 - 13 | 2314 | 86 | ||
| XM_021254674.1 | XP_021110333.1 | 11 - 12 | 2257 | 86 | ||
| XM_021254675.1 | XP_021110334.1 | 7 - 8 | 1958 | 86 | ||
| XM_021254676.1 | XP_021110335.1 | 10 - 11 | 2217 | 86 | ||

LOC101724144
Genomic Map