Assay Details
| Gene Symbol: LOC101724144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840404.2 | XP_004840461.1 | 9 - 10 | 2311 | 85 | |
| XM_021254663.1 | XP_021110322.1 | 15 - 16 | 4056 | 85 | ||
| XM_021254664.1 | XP_021110323.1 | 13 - 14 | 3264 | 85 | ||
| XM_021254665.1 | XP_021110324.1 | 13 - 14 | 3441 | 85 | ||
| XM_021254666.1 | XP_021110325.1 | 13 - 14 | 3126 | 85 | ||
| XM_021254667.1 | XP_021110326.1 | 13 - 14 | 2788 | 85 | ||
| XM_021254668.1 | XP_021110327.1 | 14 - 15 | 3984 | 85 | ||
| XM_021254669.1 | XP_021110328.1 | 14 - 15 | 3981 | 85 | ||
| XM_021254670.1 | XP_021110329.1 | 12 - 13 | 3189 | 85 | ||
| XM_021254671.1 | XP_021110330.1 | 14 - 15 | 3304 | 85 | ||
| XM_021254672.1 | XP_021110331.1 | 15 - 16 | 4024 | 85 | ||
| XM_021254673.1 | XP_021110332.1 | 14 - 15 | 2718 | 85 | ||
| XM_021254674.1 | XP_021110333.1 | 13 - 14 | 2661 | 85 | ||
| XM_021254675.1 | XP_021110334.1 | 9 - 10 | 2362 | 85 | ||
| XM_021254676.1 | XP_021110335.1 | 12 - 13 | 2621 | 85 | ||

LOC101724144
Genomic Map