Assay Details
| Gene Symbol: LOC101696935 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840505.2 | XP_004840562.1 | 7 - 8 | 1917 | 69 | |
| XM_004840506.3 | XP_004840563.1 | 7 - 8 | 2036 | 69 | ||
| XM_004840507.2 | XP_004840564.1 | 6 - 7 | 1815 | 69 | ||
| XM_021254806.1 | XP_021110465.1 | 7 - 8 | 1938 | 69 | ||
More Information
Set Membership: |
|

LOC101696935
Genomic Map