Assay Details
| Gene Symbol: Rpl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842053.2 | XP_004842110.1 | 6 - 7 | 847 | 97 | |
| XM_021256020.1 | XP_021111679.1 | 6 - 7 | 777 | 97 | ||
More Information
Set Membership: |
|

Rpl7
Genomic Map