Assay Details
| Gene Symbol: Rnf34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843844.2 | XP_004843901.2 | 4 - 5 | 1092 | 66 | |
| XM_013071790.2 | XP_012927244.2 | 4 - 5 | 1092 | 66 | ||
| XM_021257513.1 | XP_021113172.1 | 4 - 5 | 994 | 66 | ||
More Information
Set Membership: |
|

Rnf34
Genomic Map