Assay Details
| Gene Symbol: LOC101724419 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848058.3 | XP_004848115.1 | 20 - 21 | 2684 | 90 | |
| XM_021261060.1 | XP_021116719.1 | 19 - 20 | 2576 | 90 | ||
| XM_021261061.1 | XP_021116720.1 | 19 - 20 | 2231 | 90 | ||
More Information
Set Membership: |
|

LOC101724419
Genomic Map