Assay Details
| Gene Symbol: Zfp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857326.2 | XP_004857383.1 | 1 - 2 | 300 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004857326.2 transcripts.
Set Membership: |
|

Zfp3
Genomic Map