Assay Details
| Gene Symbol: Uxt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004875312.3 | XP_004875369.1 | 3 - 4 | 418 | 66 | |
| XM_004875313.3 | XP_004875370.1 | 4 - 5 | 367 | 66 | ||
More Information
Set Membership: |
|

Uxt
Genomic Map