Assay Details
| Gene Symbol: Fer | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013071121.2 | XP_012926575.1 | 5 - 6 | 761 | 73 | |
| XM_021255851.1 | XP_021111510.1 | 5 - 6 | 784 | 73 | ||
| XM_021255852.1 | XP_021111511.1 | 4 - 5 | 628 | 73 | ||
| XM_021255853.1 | XP_021111512.1 | 5 - 6 | 761 | 73 | ||
| XM_021255854.1 | XP_021111513.1 | 5 - 6 | 761 | 73 | ||
More Information
Set Membership: |
|

Fer
Genomic Map