Assay Details
| Gene Symbol: Lrrc34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013071501.2 | XP_012926955.1 | 9 - 10 | 1048 | 84 | |
| XM_021253982.1 | XP_021109641.1 | 9 - 10 | 1048 | 84 | ||
| XM_021253983.1 | XP_021109642.1 | 9 - 10 | 1045 | 84 | ||
| XM_021253987.1 | XP_021109646.1 | 9 - 10 | 1045 | 84 | ||
| XM_021253996.1 | XP_021109655.1 | 9 - 10 | 958 | 84 | ||
| XM_021253999.1 | XP_021109658.1 | 8 - 9 | 952 | 84 | ||
| XM_021254001.1 | XP_021109660.1 | 7 - 8 | 774 | 84 | ||
More Information
Set Membership: |
|

Lrrc34
Genomic Map