Assay Details
| Gene Symbol: LOC101696740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013074155.2 | XP_012929609.2 | 7 - 8 | 645 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101696740| Gene Symbol: LOC101696740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013074155.2 | XP_012929609.2 | 7 - 8 | 645 | 64 | |
Set Membership: |
|