Assay Details
| Gene Symbol: Hhla2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013075481.2 | XP_012930935.1 | 1 - 2 | 174 | 90 | |
| XM_013075484.2 | XP_012930938.1 | 1 - 2 | 174 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013075481.2, XM_013075484.2 transcripts.
Set Membership: |
|

Hhla2
Genomic Map