Assay Details
| Gene Symbol: Hhla2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013075481.2 | XP_012930935.1 | 2 - 3 | 245 | 153 | |
| XM_013075484.2 | XP_012930938.1 | 2 - 3 | 245 | 153 | ||
| XM_021264100.1 | XP_021119759.1 | 2 - 3 | 376 | 153 | ||
| XM_021264105.1 | XP_021119764.1 | 3 - 4 | 392 | 153 | ||
| XM_021264113.1 | XP_021119772.1 | 2 - 3 | 394 | 153 | ||
| XM_021264124.1 | XP_021119783.1 | 2 - 3 | 165 | 153 | ||
More Information
Set Membership: |
|

Hhla2
Genomic Map