Assay Details
| Gene Symbol: Prss48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021247890.1 | XP_021103549.1 | 4 - 5 | 802 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prss48| Gene Symbol: Prss48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021247890.1 | XP_021103549.1 | 4 - 5 | 802 | 79 | |
Set Membership: |
|