Assay Details
| Gene Symbol: Znf445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835069.3 | XP_004835126.1 | 6 - 7 | 1271 | 79 | |
| XM_021257384.1 | XP_021113043.1 | 6 - 7 | 1432 | 79 | ||
| XM_021257387.1 | XP_021113046.1 | 6 - 7 | 1397 | 79 | ||
| XM_021257392.1 | XP_021113051.1 | 6 - 7 | 1510 | 79 | ||
| XM_021257393.1 | XP_021113052.1 | 6 - 7 | 1234 | 79 | ||
More Information
Set Membership: |
|

Znf445
Genomic Map