Assay Details
| Gene Symbol: Rps21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841008.3 | XP_004841065.1 | 5 - 6 | 387 | 63 | |
| XM_004841009.3 | XP_004841066.1 | 5 - 6 | 371 | 63 | ||
More Information
Set Membership: |
|

Rps21
Genomic Map