Assay Details
| Gene Symbol: Ogfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841023.3 | XP_004841080.1 | 5 - 6 | 623 | 55 | |
| XM_004841024.3 | XP_004841081.1 | 5 - 6 | 562 | 55 | ||
| XM_021255223.1 | XP_021110882.1 | 5 - 6 | 1540 | 55 | ||
More Information
Set Membership: |
|

Ogfr
Genomic Map