Assay Details
| Gene Symbol: LOC101705246 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843107.3 | XP_004843164.2 | 2 - 3 | 909 | 110 | |
| XM_013071584.2 | XP_012927038.1 | 2 - 3 | 959 | 110 | ||
| XM_021256942.1 | XP_021112601.1 | 2 - 3 | 390 | 110 | ||
| XM_021256946.1 | XP_021112605.1 | 2 - 3 | 960 | 110 | ||
| XM_021256948.1 | XP_021112607.1 | 2 - 3 | 956 | 110 | ||
More Information
Set Membership: |
|

LOC101705246
Genomic Map