Assay Details
| Gene Symbol: Washc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844847.2 | XP_004844904.1 | 1 - 2 | 146 | 91 | |
| XM_004844848.2 | XP_004844905.1 | 1 - 2 | 122 | 91 | ||
| XM_021258409.1 | XP_021114068.1 | 1 - 2 | 122 | 91 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004844847.2, XM_004844848.2, XM_021258409.1 transcripts.
Set Membership: |
|

Washc3
Genomic Map