Assay Details
| Gene Symbol: Washc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844847.2 | XP_004844904.1 | 2 - 3 | 241 | 67 | |
| XM_004844848.2 | XP_004844905.1 | 2 - 3 | 217 | 67 | ||
More Information
Set Membership: |
|

Washc3
Genomic Map