Assay Details
| Gene Symbol: Washc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844847.2 | XP_004844904.1 | 6 - 7 | 591 | 118 | |
| XM_004844848.2 | XP_004844905.1 | 6 - 7 | 564 | 118 | ||
| XM_004844849.2 | XP_004844906.1 | 5 - 6 | 445 | 118 | ||
| XM_021258409.1 | XP_021114068.1 | 5 - 6 | 501 | 118 | ||
More Information
Set Membership: |
|

Washc3
Genomic Map