Assay Details
| Gene Symbol: Frs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845061.3 | XP_004845118.1 | 6 - 7 | 666 | 62 | |
| XM_013072332.2 | XP_012927786.1 | 3 - 4 | 397 | 62 | ||
| XM_021258546.1 | XP_021114205.1 | 7 - 8 | 765 | 62 | ||
| XM_021258547.1 | XP_021114206.1 | 5 - 6 | 625 | 62 | ||
More Information
Set Membership: |
|

Frs2
Genomic Map