Assay Details
| Gene Symbol: Csnk2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846979.2 | XP_004847036.1 | 5 - 6 | 519 | 54 | |
| XM_021260111.1 | XP_021115770.1 | 5 - 6 | 443 | 54 | ||
More Information
Set Membership: |
|

Csnk2b
Genomic Map