Assay Details
| Gene Symbol: Whamm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852859.3 | XP_004852916.1 | 9 - 10 | 2139 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Whamm| Gene Symbol: Whamm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852859.3 | XP_004852916.1 | 9 - 10 | 2139 | 61 | |
Set Membership: |
|