Assay Details
| Gene Symbol: Cdca7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857689.2 | XP_004857746.1 | 8 - 9 | 1310 | 66 | |
| XM_004857691.2 | XP_004857748.1 | 7 - 8 | 1072 | 66 | ||
| XM_013076937.2 | XP_012932391.1 | 9 - 10 | 1366 | 66 | ||
| XM_021238609.1 | XP_021094268.1 | 9 - 10 | 1347 | 66 | ||
| XM_021238610.1 | XP_021094269.1 | 9 - 10 | 1344 | 66 | ||
| XM_021238611.1 | XP_021094270.1 | 9 - 10 | 1317 | 66 | ||
| XM_021238612.1 | XP_021094271.1 | 8 - 9 | 1293 | 66 | ||
| XM_021238613.1 | XP_021094272.1 | 10 - 11 | 1458 | 66 | ||
| XM_021238614.1 | XP_021094273.1 | 7 - 8 | 1056 | 66 | ||
| XM_021238615.1 | XP_021094274.1 | 7 - 8 | 1014 | 66 | ||
More Information
Set Membership: |
|

Cdca7
Genomic Map