Assay Details
| Gene Symbol: Nr5a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860118.3 | XP_004860175.1 | 6 - 7 | 1213 | 70 | |
| XM_004860119.3 | XP_004860176.1 | 4 - 5 | 1190 | 70 | ||
| XM_004860120.2 | XP_004860177.1 | 5 - 6 | 1173 | 70 | ||
| XM_004860121.3 | XP_004860178.1 | 4 - 5 | 1145 | 70 | ||
| XM_021240654.1 | XP_021096313.1 | 5 - 6 | 1076 | 70 | ||
| XM_021240655.1 | XP_021096314.1 | 4 - 5 | 1370 | 70 | ||
More Information
Set Membership: |
|

Nr5a2
Genomic Map